Learn More
Cayman Chemical Annaosanchun 10mg
SDP
Supplier:  Cayman Chemical 3757210 MG
Encompass_Preferred
A synthetic sterol and neuroprotective agent; increases the viability of, and the levels of ATP and the mitochondrial membrane potential in, hypoxia-exposed and reoxygenated primary rat cortical neurons at 0.1, 0.3, and 1 µM; prevents glutamate-induced increases in intracellular acidification, lactate levels, and glycolysis rate, as well as inhibits glutamate-induced apoptosis, in primary rat cerebral granule neurons at 10 µM; reduces neurological and neuromuscular deficits and infarct volume in a rat model of MCAO-induced ischemia-reperfusion injury at 0.5, 2, or 10 mg/kg
Catalog No. 50-291-6106