Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Annexin A1/ANXA1 Antibody (YA832) | Affinity purified | 39 kDa | 50 UL
SDP

Supplier:  Medchemexpress LLC HYP8054450UL

Encompass_Preferred

This Annexin A1/ANXA1 Antibody (YA832) is a mouse-derived, non-conjugated IgG1 monoclonal antibody targeting Annexin A1/ANXA1. It is supplied as a liquid formulation and functions in innate immune response, anti-inflammatory activity, and T-cell regulation.

  • Host: Mouse
  • Clonality: Monoclonal
  • Molecular weight: 39 kDa
  • Species reactivity: Human, mouse, rat, monkey
  • Immunogen: Synthetic peptide for human Annexin A1 beta
  • Application: Western blot (1:500-1:1000)
  • Purity: Affinity purified
  • Conjugation: Non-conjugated
  • Isotype: IgG1
  • Formulation: PBS (pH 7.3), 50% glycerol, 0.5% BSA, 0.02% sodium azide
  • Storage: -20°C for 1 year; avoid freeze/thaw cycles
  • Shipping: Blue ice
  • Functions: Innate immune response, anti-inflammatory, T-cell regulation, phagocytosis, chemotaxis

Catalog No. 50-003-95992


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.