Learn More
Medchemexpress LLC Annexin A5/ANXA5 protein, mouse (His, Myc) | >95.0% | 500UG
SDP
Supplier:  Medchemexpress LLC HYP702573500UG
Encompass_Preferred
Recombinant mouse Annexin A5 is supplied as a lyophilized His- and Myc-tagged protein for use in coagulation, apoptosis, and biochemical studies. The product is expressed in Escherichia coli and is provided with supporting datasheet and certificate of analysis for experimental reproducibility.
  • Expressed in Escherichia coli for efficient production.
  • N-terminal 10xHis and C-terminal Myc tags for detection and purification.
  • >95% purity by reducing SDS-PAGE.
  • Molecular weight approximately 38-41 kDa.
  • Lyophilized formulation with stabilizing excipient for storage.
  • Available in multiple sizes including a 500 μg vial.
Catalog No. 50-001-76019