Learn More
TARGETMOL CHEMICALS INC Anpirtoline hydrochloride 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T225765MG
Encompass_Preferred
Also available in 1 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. Anpirtoline hydrochloride (D-16949 hydrochloride) is a novel and potent 5-HT1B receptor agonist and 5-HT3 receptor antagonist with anti-injurious, analgesic, and antidepressant activity, and is used in the study of movement disorders. Purity 100%
Catalog No. 50-287-7734