Learn More
Medchemexpress LLC Anpirtoline hydrochloride | 99201-87-3 | 99.9% | 265.20 g/mol | C10H14Cl2N2S | 25 MG
SDP
Supplier:  Medchemexpress LLC HY10110025MG
Encompass_Preferred
Anpirtoline hydrochloride is the hydrochloride salt of anpirtoline, a potent, centrally acting 5-HT1B receptor agonist used in neuroscience research. It displays activity across serotonin receptor subtypes and is supplied as a high-purity research reagent for pharmacology and biochemical assays.
  • Potent 5-HT1B receptor agonist (Ki ≈ 28 nM).
  • Also shows activity at 5-HT1A and 5-HT2 receptors (Ki ≈ 150 nM and 1.49 μM).
  • Purity 99.86% for reproducible experimental results.
  • Suitable for in vitro and in vivo pharmacology studies.
Catalog No. 50-002-13293