Learn More
BIOHIPPO AnRE-TAL-Puro LV
SDP
Supplier:  BIOHIPPO BHV194000413
Encompass
AnRE-TAL-Puro is a transcription factor reporter lentiviral vector designed to monitor Androgen receptor (AR) signaling pathway in living cells This construct delivers a Firefly Luc reporter cassette together with a Puromycin selection marker for generation of stable cell pools The vector is provided as a ready to use Lentivirus preparation pseudotyped with VSV-G for efficient gene delivery and stable genomic integration Each vial provides a nominal titer of 2x10E6 transducing units TU Typical applications include Monitor AR signaling pathway activity in mammalian cells screen for activators or inhibitors of the AR signaling pathway The fluorescent reporter enables convenient readout using flow cytometry fluorescence microscopy etc The system is suitable for use with Human/mouse and can support assay development high content screening and mechanism of action studies This product is intended for research use only and is not for diagnostic or therapeutic procedures
Catalog No. 50-003-31190