Learn More
Medchemexpress LLC ANS (8-Anilino-1-naphthalenesulfonic acid) | 82-76-8 | 98.01% | 5 G
SDP
Supplier:  Medchemexpress LLC HYD08965G
Encompass_Preferred
ANS (8-Anilino-1-naphthalenesulfonic acid) is a competitive inhibitor targeting thyroxine binding globulin (TBG). It is used in radioimmunoassay to enhance detection sensitivity by displacing bound triiodothyronine (T3), releasing free T3 for antibody recognition. Additionally, it functions as a fluorescent probe by binding to hydrophobic regions of proteins to monitor conformational dynamics through changes in fluorescent signals.
  • Utilized to enhance radioimmunoassay detection sensitivity
  • Acts as a fluorescent probe for protein conformational dynamics
  • Widely applied in biochemical research
  • Used in the development of antibacterial materials
Catalog No. 50-004-10691