Learn More
Medchemexpress LLC ANS ammonium | 28836-03-5 | 99.9% | 100 G
Supplier: Medchemexpress LLC HYD0896A100G
ANS ammonium is a potent antibacterial agent and a textile dye. It can be used as a fluorescence probe and blocks the binding of triiodothyronine to thyroxine binding globulin in radioimmunoassay of triiodothyronine.
- Potent antibacterial agent
- Textile dye
- Can be used as a fluorescence probe
- Blocks the binding of triiodothyronine to thyroxine binding globulin in radioimmunoassay of triiodothyronine
- Appearance: Solid
- Color: Off-white to gray