Learn More
Enterprise Technology Solutions Ansell Healthcare Products Gloves,Nitrile,PF,Textured,5.9mil,12"Large,M,50/BX,10/CT,PE
SDP
Supplier:  Enterprise Technology Solutions ANS50602
Encompass_Preferred
Purple Nitrile-Xtra Exam Gloves are made of highly durable nitrile polymer and guard against incidental splashes. Gloves are powder-free, silicon-free and latex-free. Textured fingertips improve grip and dexterity to better perform sensitive tasks. Because gloves are ambidextrous, you can quickly grab a single glove for either hand. Beaded cuffs make gloves easier to don and prevent roll-down. 5.9 mil thick gloves are available in various sizes. Exam gloves are cleared for use in chemotherapy and can be diverted from the landfill through The Right Cycle Program.
Catalog No. 50-008-28589