Learn More
Ansell™ Vantage™ Cut-Resistant Gloves
Supplier:  Ansell™ 245717
Catalog No. 19-920-1778
Only null left
Add to Cart
This item is not returnable. View return policy
Description
Description
Revolutionary engineered yarns provide increased cut protection without the bulk. Ideal for work environments where sharp parts and materials are handled.
  • Blended fiber, lightweight construction provides all-day comfort
  • Contour design offers flexibility for reduced hand fatigue
  • Medium weight, 10-gauge yarn with 100% Kevlar™ reinforced thumb crotch
Compliance
ANSI™ Level 4 cut protection
TRUSTED_SUSTAINABILITY
Product Suggestions
Product Suggestions