Learn More
Medchemexpress LLC Ansornitinib | 1448874-96-1 | 98.2% | 540.61 | 100 MG
SDP
Supplier:  Medchemexpress LLC HY147243100MG
Encompass_Preferred
Ansornitinib is an orally active dual kinase inhibitor that inhibits platelet-derived growth factor receptor (PDGFR) and vascular endothelial growth factor receptor (VEGFR2). It can be used as an antifibrotic agent in lung, liver, kidney, and gastrointestinal fibrotic diseases.
  • Orally active dual kinase inhibitor
  • Inhibits platelet-derived growth factor receptor (PDGFR)
  • Inhibits vascular endothelial growth factor receptor (VEGFR2)
  • Functions as an antifibrotic agent
  • Significantly inhibits phosphorylation of PDGFRβ and KDR
  • Reduces inflammation and fibrosis markers
  • Shown to reduce lung fibrosis in animal models
  • Improves colonic histology in animal models
Catalog No. 50-003-61981