Learn More
ABclonal Technology ANT2/SLC25A5 Rabbit mAb
SDP
Supplier:  ABclonal Technology A23411100UL
Encompass_Preferred
This gene is a member of the mitochondrial carrier subfamily of solute carrier protein genes The product of this gene functions as a gated pore that translocates ADP from the cytoplasm into the mitochondrial matrix and ATP from the mitochondrial matrix into the cytoplasm The protein forms a homodimer embedded in the inner mitochondria membrane Suppressed expression of this gene has been shown to induce apoptosis and inhibit tumor growth The human genome contains several non-transcribed pseudogenes of this gene
Catalog No. 50-000-06397