Learn More
Medchemexpress LLC Antalarmin hydrochloride | 220953-69-5 | MFCD05664721 | 99.9% | 415.01 g/mol | C24H35ClN4 | 10 MG
SDP
Supplier:  Medchemexpress LLC HY10337710MG
Encompass_Preferred
Antalarmin hydrochloride is a non-peptide corticotropin-releasing factor receptor 1 (CRF1/CRHR1) antagonist supplied for research use. It is used in preclinical studies to inhibit CRF-mediated signaling and to investigate stress-, anxiety-, and HPA axis-related physiological and inflammatory responses.
  • Non-peptide CRF1 (CRHR1) antagonist with high affinity.
  • Used in preclinical studies of stress, anxiety, and HPA axis modulation.
  • High purity solid, white to light yellow, suitable for analytical and in vivo experiments.
  • Available in small quantity packs and as DMSO solution aliquots for convenience.
  • Recommended storage at -20°C; protect from moisture and avoid repeated freeze-thaw cycles.
Catalog No. 50-002-45179