Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Antalarmin (hydrochloride) | 220953-69-5 | MFCD05664721 | 99.9% | 415.01 g/mol | C24H35ClN4 | 25 MG
SDP

Supplier:  Medchemexpress LLC HY10337725MG

Encompass_Preferred

Antalarmin hydrochloride is a research-grade, non-peptide corticotropin-releasing hormone receptor 1 (CRHR1) antagonist supplied as a solid for preclinical pharmacology and biochemical studies. Intended for laboratory research use only.

  • High purity (≈99.9%).
  • Molecular weight 415.01 g/mol.
  • Molecular formula C24H35ClN4.
  • Appearance: solid, white to light yellow.
  • Storage: sealed at -20°C; in solution: -80°C for 6 months, -20°C for 1 month.
  • Supplied with data sheet, certificate of analysis, and safety data sheet.

Catalog No. 50-002-45177


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.