Learn More
Medchemexpress LLC Antazoline hydrochloride | 2508-72-7 | 99.6% | 1 G
SDP
Supplier:  Medchemexpress LLC HYB10671G
Encompass_Preferred
Antazoline hydrochloride is a histamine H1 receptor antagonist known for its anticholinergic and antiviral properties. It works by reversibly competing for histamine receptor sites, thereby blocking and inhibiting the H1 receptor to produce an antiallergic effect. This compound can effectively reduce HBV DNA in the extracellular supernatant in a dose-dependent manner and has also demonstrated an anti-arrhythmic effect in acute myocardial infarctions, making it suitable for research in cardiovascular diseases and HBV.
  • Acts as a histamine H1 receptor antagonist
  • Exhibits anticholinergic and antiviral properties
  • Effectively reduces HBV DNA in HepAD38 and Huh7 cells
  • Demonstrates anti-arrhythmic effects
  • Suitable for research in cardiovascular diseases and HBV
Catalog No. 50-004-14037