Learn More
Supply Solutions Sge - 25 Ul 23 Ga Gas Tight Fixed Needle Syringe; Gc Autosampler Syringe; Ctc Pal; Syringe Code: 25F-C/T-Gt-5/0.63C; Od (Mmm) 0.63
Supplier:  Supply Solutions SGE 003987
Encompass
SGE - 25 uL 23 GA Gas Tight Fixed Needle Syringe; GC Autosampler syringe; CTC PAL; Syringe Code: 25F-C/T-GT-5/0.63C; OD (MMM) 0.63; Needle Length (MM) 50 gas tight; Tip Type: Cone; Replacement Plunger: 031817; Volume: 25uL; Gauge 23; For Use With Syringe 003987; For Use With (Application) IN VITRO use only. For lab analyzers only. Not for human use.; For Use With (Equipment) For 25uL fixed needle gas tight syringe for CTC/Carlo Erba/ Fisons Autosampler; Material PTFE tip
Catalog No. NC0010223