Learn More
Medchemexpress LLC Antennapedia peptide (penetratin peptide) | 188842-14-0 | 98.4% | 2246.73 | 5 MG
SDP
Supplier:  Medchemexpress LLC HYP03075MG
Encompass_Preferred
Antennapedia peptide (Penetratin peptide) is a 16 amino acid peptide derived from the 60 amino acid long homeodomain of the Drosophila transcription factor Antennapedia. As a member of the cell-penetrating peptides family, it activates caspase-dependent apoptosis and has been shown to have radiation-sensitising effects on tumor cells.
  • Derived from Drosophila transcription factor Antennapedia
  • 16 amino acid peptide
  • Member of cell-penetrating peptides family
  • Activates caspase-dependent apoptosis
  • Has radiation-sensitising effects
  • Increases levels of cleaved caspase-3
Catalog No. 50-004-24455