Learn More
Genetex Inc Anterior Gradient 2 Monoclonal Antibody [HMV325] HistoMAX™ (GTX640557) GeneTex - IHC-Optimized Antibody
SDP
Supplier:  Genetex Inc GTX640557
Encompass_Preferred
The Anterior Gradient 2 antibody [HMV325] HistoMAX™ (GTX640557) is a Rabbit Monoclonal antibody in the GeneTex HistoMAX portfolio. The antibody is extensively evaluated for IHC-P using formalin-fixed human normal tissues (more than 70 different sections) and cancer tissue arrays to guarantee specificity and best-in-class performance. Information regarding both manual and autostainer staining protocols are available.Figure Legend: IHC-P analysis of human stomach tissue using GTX640557 Anterior Gradient 2 antibody [HMV325] HistoMAX. Strong predominantly cytoplasmic anterior gradient 2 staining of all epithelial cells except parietal cells which only show a faint staining.
Catalog No. NC3669337