Learn More
AdipoGen Anthracene-9,10-dipropionic acid disodium salt
SDP
Supplier:  AdipoGen CDXA0116M010
Encompass_Preferred
Anthracene-9,10-dipropionic acid disodium salt, AdipoGen Life Sciences. Chemical. CAS: 82767-90-6. Formula: C20H16Na2O4. MW: 366.32. Synthetic. Water-?soluble naphthalene as singlet oxygen sensitizers. Self-?assembly building block.
Catalog No. 50-596-560