Learn More
STEMCELL Technologies Doxorubicin (Hydrochloride), Size: 25 mg
SDP
Supplier:  STEMCELL Technologies 1000559
Encompass
Doxorubicin is an anthracycline antibiotic that intercalates DNA and inhibits DNA topoisomerase II (Burridge et al.; Lorenzo et al.). Doxorubicin exhibits cardiotoxic effects that involve generating reactive oxygen species, inhibiting DNA topoisomerase II, and releasing calcium in the sarcoplasmic reticulum (Burridge et al.). This product is supplied as the hydrochloride salt of the molecule.CANCER RESEARCH· Induces apoptosis in human endothelial cells (Lorenzo et al.).
Catalog No. 50-206-9367