Learn More
AdipoGen Anthrance A
SDP
Supplier:  AdipoGen CDXA0585M100
Encompass_Preferred
Chemical. CAS 185318-74-5. Formula C20H8F4O5. MW 404.27. Fluorescent probe/dye with high absorbance and favorable resistance to photobleaching. Spectral Data Absorption/Emission=508/527nm.
Catalog No. 50-256-1731