Learn More
Medchemexpress LLC Anthraquinone-2-carboxylic acid | 117-78-2 | MFCD00001231 | 98.0% | 252.22 g/mol | C15H8O4 | 1 ML

Supplier: Medchemexpress LLC HYW03175710MM/1ML
Anthraquinone-2-carboxylic acid is an anthraquinone derivative isolated from Brazilian taheebo with reported anti-inflammatory and antinociceptive activity. It is supplied for research use in powder form or as a 10 mM solution in DMSO, and is intended for in vitro applications when handled and stored per recommended conditions.
- CAS number 117-78-2.
- Purity 98.0%.
- Molecular formula C15H8O4; molecular weight 252.22 g/mol.
- Available as powder and as a 10 mM solution in DMSO (1 mL).
- Soluble in DMSO at 25 mg/mL (≈99.12 mM); ultrasonic assistance recommended.
- Powder storage: -20°C (3 years) or 4°C (2 years).
- In-solvent storage: -80°C (6 months) or -20°C (1 month).
- For research use only.
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.