Learn More
PROGEN BIOTECH INC anti-AAV VP1/VP2/VP3 mouse monoclonal, B1, liquid, purified, sample, 200ul
Supplier:  PROGEN BIOTECH INC 690058S
Encompass
-Mouse monoclonal. -Suitable for affinity chromatography, dot blot, ICC/IF, IP and WB. -Reacts with denatured VP1, VP2, VP3 proteins of AAV1, 2, 3, 5, 6, 7, 8, 9, rh10, DJ. -Isotype: IgG1
Catalog No. NC3789639