Learn More
ABclonal Technology ACTB Monoclonal Antibody,100UL,Validated in: WB,IHC,IF
SDP
Supplier:  ABclonal Technology AC004
Encompass_Preferred
Immunogen: Fusion protein of human β-Actin•Host Species: Mouse•Isotype: IgG•Species Reactivity:Human,Mouse,Rat•Validated Applications:WB,IF•Swiss Prot: P60709•Gene ID:60•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-164-9225