Learn More
Santa Cruz Biotechnology Anti-beta-Actin Antibody (AC-15), 100 ug/mL
Supplier:  Santa Cruz Biotechnology SC69879
Encompass
Recommended for detection of beta-Actin of broad species origin by WB, IP and IF; not recommended for detection of adult cardiac and skeletal muscle actins or detection in Dictyostelium discoideum.
Catalog No. NC9666552