Learn More
Abcam Anti-ADA antibody [EPR25429-117]
Supplier:  Abcam AB300050
Encompass_Preferred
Rabbit Recombinant Monoclonal ADA antibody. Suitable for WB, IHC-P, ICC/IF, IP and reacts with Human samples.
Catalog No. NC3855936