Learn More
Advanced ImmunoChemical, Inc. Monoclonal Mouse Anti-Human Adiponectin, 0.2mg
Supplier:  Advanced ImmunoChemical, Inc. 1ADP0.2 MG
Encompass_Preferred
This item is manufactured by HyTest, Ltd and distributed by Advanced ImmunoChemical, Inc. Monoclonal Antibodies / Metabolic Syndrome/ Adiponectin. MAbs: Adn23, Adn27, Adn36, Adn63, Adn94, Adn214, Adn222, Adn279, Adn305. Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with human recombinant adiponectin from E. coli (MAbs Adn23) or with native human Monoclonal Antibodies. Metabolic Syndrome. Monoclonal Mouse Anti-Adiponectin (MAbs Adn27, Adn36, Adn63, Adn94, Adn214, Adn222, Adn279 and Adn305). Specificity: Human serum adiponectin.
Catalog No. 50-153-9824