Learn More
Sino Biological Anti-Alkaline phosphatase/APLI Antibody, Rabbit Polyclonal 100 µL
Supplier:  Sino Biological 13225T24100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant Human Alkaline phosphatase/APLI (rh Alkaline phosphatase/APLI; Catalog#13225-H08H; P09923; Met1-Asp503). Alkaline phosphatase/APLI specific IgG was purified by Human Alkaline phosphatase/APLI affinity chromatography.
Catalog No. 50-007-48988