Learn More
Selleck Chemical LLC Anti-Amyloid Beta-A2813-1MG-25
SDP
Supplier:  Selleck Chemical LLC A28131MG25
Encompass_Preferred
Anti-Amyloid Beta is a recombinant monoclonal antibody in a single chain fragment variable (scFv) against amyloid beta has the potential to be used in Alzheimer s disease (AD) research MW 145 14 KD
Catalog No. 50-313-5356
Add to Cart
Request a Quote