Learn More
Rockland Immunochemicals Anti-Amyloid Oligomers (A11) (RABBIT) Antibody - 200-401-E88
SDP
Supplier:  Rockland Immunochemicals 200401E88
Encompass
Amyloid monomeric proteins can sometimes oligomerize into destructive amyloid fibrils. Amyloidogenic conformations of non-disease related proteins can be created by partial protein misfolding or denaturation. Many degenerative diseases are known to be related to the accumulation of misfolded proteins as amyloid fibres. These include the amyloid-β peptide plaques and tau neurofibrillary tangles in senile plaques of Alzheimer’s symptomology, the deposition of α-synuclein in the Lewy bodies of Parkinson’s disease, and accumulation of polyglutamine-containing aggregates in Huntington’s disease. Anti-Amyloid Oligomers A11 Antibody is useful for researchers interested in Neuroscience research.
Catalog No. 50-105-7647