Learn More
Medchemexpress LLC Anti-ANGPTL4 Antibod 5mg | 5mg
SDP
Supplier:  Medchemexpress LLC HYP9903355MG
Encompass_Preferred
Anti-ANGPTL4 Antibody is a CHO expressed human antibody targeting ANGPTL4 Anti-ANGPTL4 Antibody has a huIgG1-type heavy chain and a hu -type light chain with a predicted molecular weight (MW) of 150 kDa An isotype control for Anti-ANGPTL4 Antibody can be found in Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-24368