Learn More
ECM Bioscience Anti-Phosphotyrosine Mouse Monoclonal Antibody, ECM Biosciences
Supplier:  ECM Bioscience PM3751
Encompass_Preferred
Clone M375 was generated from tyrosine phosphorylated proteins purified from pervanadate treated A431 cells. Accession #. This antibody detects phosphotyrosine-containing proteins on SDS-PAGE immunoblots of EGF treated A431 cells, as well as pervanadate treated A431, HeLa, Jurkat, and endothelial cells. Applications: WB, E, IP, ICC; Species Reactivity: Hu, Rt, Ms, Ck; Size = 100 μl; Buffer = Mouse monoclonal antibody purified with protein A chromatography is supplied in 100µl phosphate-buffered saline, 50% glycerol, 1 mg/ml BSA, and 0.05% sodium azide. Store at –20°C. Stable for 1 year. Other Names = phosphotyrosine, phoshoserine, phoshothreonine
Catalog No. 50-206-8797