Learn More
Medchemexpress LLC Anti-B7-H4 VTCN1 Ant 5mg | 5mg
SDP
Supplier:  Medchemexpress LLC HYP9903415MG
Encompass_Preferred
Anti-B7-H4/VTCN1 Antibody is a CHO-expressed human antibody that targets B7-H4/VTCN1 The Anti-B7-H4/VTCN1 Antibody contains a huIgG1 heavy chain and a hu light chain with a predicted molecular weight (MW) of 145 kDa The isotype control for Anti-B7-H4/VTCN1 Antibody can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-25492