Learn More
Immunology Consultants anti-BVDV Antibody (Mouse - Clone #104) - Monoclonal
SDP
Supplier:  Immunology Consultants MBVD10A104
Encompass_Preferred
This mouse monoclonal antibody provides sensitive, specific detection of Bovine Viral Diarrhea Virus (BVDV) antigens. Clone 104 performs reliably in ELISA, Western blotting, and antigen‑capture assays, offering strong affinity and low nonspecific binding. It is well suited for virology research, vaccine evaluation, and surveillance of BVDV infection in cattle. MBVD‑10A‑104 delivers consistent, high‑clarity performance across repeated assays and diverse sample matrices.
Catalog No. 50-004-35367