Learn More
Immunology Consultants anti-BVDV Antibody (Mouse - Clone #105) - Monoclonal
SDP
Supplier:  Immunology Consultants MBVD10A105
Encompass_Preferred
Clone 105 targets a distinct BVDV epitope, enabling precise detection in immunoassays. Its strong affinity and clean signal support ELISA, Western blotting, and antigen‑capture workflows. MBVD‑10A‑105 is ideal for diagnostic development, vaccine research, and monitoring BVDV exposure in livestock populations. Its reproducible performance ensures dependable antigen detection across varied sample types and assay conditions.
Catalog No. 50-004-35500