Learn More
Immunology Consultants Monoclonal Mouse anti-C-MYC Antibody Clone#4E10, 1.0 mg
SDP
Supplier:  Immunology Consultants MMYC45A4E10
Encompass_Preferred
The C-Myc tag (EQKLISEEDL-tag) has been widely used in recombinant protein expression and production. It allows simple and efficient affinity purification of the tagged protein using Myc-tag specific antibody conjugated to agarose-beads.  This antibody is suitable for ELISA, Blotting, IF and IP applications.
Catalog No. NC2734271