Learn More
Advanced ImmunoChemical, Inc. ANTI-CA72-4 MAB
Supplier:  Advanced ImmunoChemical, Inc. 2CA721 MG
Encompass_Preferred
MONOCLONAL ANTIBODIES/Monoclonal mouse anti-CA72-4. MAbs: 7C1,1C2,2C3.Hybridoma clones have been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with native CA72-4. MAb isotypes: IgG1 for MAbs 7C1,1C2,2C3Applications:Detection of CA72-4 antigen. Recommended pairs for sandwich immunoassay (capture – detection):1C2 – 7C12C3 – 7C1All MAbs are working in Western blotting. Purifications: Ion exchange chromatographyPresentationPBS, pH 7.4, 0.09 % sodium azide (NaN₃)Storage Conditions:+4 °C (+2 … +8 °C allowed)
Catalog No. NC3566212