Learn More
Immunology Consultants anti-Canine Heartworm Antibody (Mouse - Clone #104) - Monoclonal
SDP
Supplier:  Immunology Consultants MHRT65A104
Encompass_Preferred
Clone 104 provides high‑specificity detection of canine heartworm antigens, supporting ELISA, Western blotting, and antigen‑capture assay development. Its strong affinity and clean signal enable sensitive detection in serum, plasma, and purified antigen preparations. MHRT‑65A‑104 is ideal for diagnostic development, parasite research, and monitoring heartworm exposure in canine populations. Its consistent lot‑to‑lot performance ensures reliable detection across repeated assays.
Catalog No. 50-004-35393