Learn More
Immunology Consultants anti-Canine Heartworm Antibody (Rabbit) - Affinity Purified
SDP
Supplier:  Immunology Consultants RHRT65A
Encompass_Preferred
This affinity‑purified rabbit anti‑canine heartworm antibody provides highly specific recognition of Dirofilaria immitis antigens, supporting sensitive detection in diagnostic assay development and research applications. Purification through antigen‑specific chromatography ensures low background and strong signal performance in ELISA, Western blotting, and immunoassay formats. Manufactured by Immunology Consultants, this 1 mg preparation offers consistent binding characteristics suitable for laboratories studying heartworm infection, parasite biology, or veterinary diagnostic workflows. Its clean profile and robust affinity make it a dependable reagent for detecting heartworm‑associated proteins in both purified and complex biological samples
Catalog No. 50-004-35405