Learn More
AdipoGen ANTI-CASPASE-8 (MOUSE), (100UG
SDP
Supplier:  AdipoGen AG20T0138C100
Encompass_Preferred
anti-Caspase-8 (mouse), mAb (3B10), AdipoGen Life Sciences. Monoclonal Antibody. Recognizes the p20 subunit of mouse caspase-8. Detects bands of ~55kDa (full-length caspase-8) and ~18kDa (apoptosis-induced cleavage fragment) by Western blot. Does not cross-react with human caspase-8. Isotype: Rat IgG1kappa. Clone: 3B10. Applications: ELISA, FACS, ICC, IHC, WB. Liquid. In PBS containing 0.02% sodium azide. Procaspase-8 belongs to the family of caspases. Binding of FasL to Fas leads to formation of a receptor complex at the cellular membrane, which was named DISC. The DISC consists of oligomerized receptors, the DD-containing adaptor molecule FADD, procaspase-8, procaspase-10 and c-FLIP. The DISC structure provides a platform for the oligomerization of procaspase-8 that allows two procaspase-8 homodimers to be in the close proximity leading to the initial activation of procaspase-8. At the first cleavage step, the N-terminal p43/p41 and the C-terminal p30 cleavage products are generated.
Catalog No. 50-101-5700