Learn More
Invivogen InvivoGen Anti-CD20 Monoclonal Antibody, Targets Human CD20, Chimeric Construct, 3 x 100 µg
SDP
Supplier:  Invivogen HCD20MAB103
Encompass
InvivoGen Anti-CD20 (Rituximab biosimilar) is a human IgG1 monoclonal antibody that targets the human CD20 antigen, facilitating cell destruction through apoptosis, complement activation, and cell-mediated cytotoxicity. Ideal for scientific research, it is produced in CHO cells with thorough functional testing verified by flow cytometry. Its azide-free, lyophilized formulation ensures stability and ease of use.
Catalog No. NC3071631