Learn More
Selleck Chemical LLC Anti-CD47-A2637-1MG-5
SDP
Supplier:  Selleck Chemical LLC A26371MG 5PC
Encompass_Preferred
Anti-CD47 is a humanized monoclonal antibody (mAb) targeting CD47 It can be used for the research of hematologic malignancies and solid tumors MW 150 KD
Catalog No. 50-313-8416