Learn More
Selleck Chemical LLC Anti-CD47-InVivo-A2140-5MG-20
SDP
Supplier:  Selleck Chemical LLC A21405MG 20PC
Encompass_Preferred
Anti-mouse/human/rat CD47-InVivo reacts with mouse CD47 otherwise known as integrin-associated protein CD47 is an approximately 50 kDa glycosylated five transmembrane protein that is ubiquitously expressed by both hematopoietic cells such as T and B lymphocytes monocytes platelets and erythrocytes and non-hematopoietic cells
Catalog No. 50-313-8013