Learn More
FUJIFILM BIOSCIENCES INC Anti CD81 Rat Monoclonal Antib
Supplier:  FUJIFILM BIOSCIENCES INC 01028223
Encompass_Preferred
CD81 is 26 kDa cell surface protein and is known as TAPA-1 Target of the Antiproliferative Antibody 1 and Tetraspanin-28 Tspan-28 CD81 interacts directly with immunoglobulin superfamily member 8 IGSF8 CD316 and CD36 It also forms a signal transduction complex with CD19 CD21 and Leu-13 CD225 on the surface of B cell In addition on T cells CD81 associates with CD4 and CD8 and provides a costimulatory signal with CD3 This antibody is established against human CD81 with Rat It specifically recognize human CD81 and very slightly bovine CD81 It does not recognize mouse and rat CD81 High specificity to human CD81
Catalog No. 50-004-50152