Learn More
FUJIFILM BIOSCIENCES INC Anti CD9 Rat Monoclonal Antibo
Supplier:  FUJIFILM BIOSCIENCES INC 01928173
Encompass_Preferred
CD9 is a cell surface glycoprotein which belongs to the tetraspanin superfamily TM4SF and is found on the surface of exosomes This is known to form a complex with integrins and other TM4SF can modulate cell adhesion and migration Also this induces platelet activation and aggregation CD9 is also found on the surface of exosomes This product is rat monoclonal antibody that recognizes CD9 Specific for CD9 antigen
Catalog No. 50-004-50171