Learn More
Medchemexpress LLC Anti-CDCP1 CD318 Ant 25mg | 25mg
SDP
Supplier:  Medchemexpress LLC HYP99036125MG
Encompass_Preferred
The Anti-CDCP1/CD318 Antibody (38 E11) is a CHO-expressed human antibody targeting CDCP1/CD318 The Anti-CDCP1/CD318 Antibody (38 E11) contains a huIgG1 heavy chain and a hu light chain with a predicted molecular weight (MW) of 150 kDa The isotype control for Anti-CDCP1/CD318 Antibody (38 E11) can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-24279