Learn More
Medchemexpress LLC Anti-CDH1 E-cadherin 1mg | 1mg
SDP
Supplier:  Medchemexpress LLC HYP9903621MG
Encompass_Preferred
Anti-CDH1/E-cadherin/CD324 Antibody is a humanized antibody expressed in CHO targeting CDH1/E-cadherin/CD324 The Anti-CDH1/E-cadherin/CD324 Antibody contains a huIgG1 heavy chain and a hu light chain with a predicted molecular weight (MW) of 146 6 kDa The isotype control for Anti-CDH1/E-cadherin/CD324 Antibody can be referenced as Human IgG1 kappa Isotype Control (HY-P99001)
Catalog No. 50-003-25267