Learn More
Biogenex Laboratories Anti-CDX-2, Clone CDX2-88
SDP
Supplier:  Biogenex Laboratories MU392A5UC
Encompass
CDX2, a member of the caudal-related homeobox family, is an intestine-specific transcription factor that regulates both proliferation and differentiation in intestinal epithelial cells. It plays an important role in triggering cells towards the phenotype of differentiated villus enterocytes as well as in the maintenance of the phenotype. Clone CDX2-88 reacts with a conserved epitope of the 40kD CDX2 protein localized in the nucleus. It exclusively marks nuclei of colonic epithelial cells and colorectal cancers on formalin-fixed, paraffin-embedded tissue sections.
Catalog No. NC1083496