Learn More
Crystal Chem Inc Anti-CHO-HCP Antibody Type A, 0.5mg
SDP
Supplier:  Crystal Chem Inc IP111
Encompass
CHO 360-HCP Antibody, Type A, from total HCP and goat. Suitable for ELISA kits or Western blotting.Crystal Chem, in partnership with German manufacturer BioGenes, has recently launched a new line of generic HCP ELISA kits for CHO cells which far outperform competitor kits for detection of host cell proteins. Unlike traditional one-size-fits-all approaches, we provide four assays, each using a different anti-HCP antibody, so you can try multiple antibodies to find the one that works best for your process. 360-HCP ELISA has demonstrated better HCP recovery, better dilution linearity, and higher inter-assay precision than competitor’s kits.
Catalog No. 50-194-7934