Learn More
ABclonal Technology COL3A1 Polyclonal Antibody, 100UL, Validated in:WB
SDP
Supplier:  ABclonal Technology A3795
Encompass_Preferred
Immunogen: A synthetic peptide corresponding to a sequence within amino acids 1200-1300 of human COL3A1 (NP_000081.2).•Host Species: Rabbit•Isotype: IgG•Species Reactivity:Human,Mouse•Validated Applications:WB,IF•Swiss Prot: P02461•Gene ID:1281•Purification Method:Affinity purification•Storage: Store at -20℃. Avoid freeze / thaw cycles.|Buffer: PBS with 0.02% sodium azide,50% glycerol,pH7.3.This product is available in 20UL, 50UL, 100UL, and 200UL. Please contact customer service for pricing and availability on alternative sizes.
Catalog No. 50-130-7580